Tween® 20 (Polysorbate)
Dodavatel: Lach-ner S.R.O.
40161FT0G01001
40161FT0G05001
LACH40161FT0G01001EA
0
CZK
LACH40161FT0G01001
LACH40161FT0G05001
Tween® 20 (Polysorbate)
Tween® 20 (Polysorbate)
Vzorec:
C₅₈H₁₁₄O₂₆ Hustota: 1,1 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00165986 CAS číslo: 9005-64-5 EINECS: 500-018-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...