Disulfid didraselný
Dodavatel: Lach-ner S.R.O.
|
Nebezpečí
|
Synonyms:
Dipotassium disulphite, Potassium pyrosulphite, Potassium metabisulphite, di-Potassium disulphite
30023AP0G0500
30023AP0G05001
30023AP0G1000
LACH30023AP0G0500EA
586
CZK
LACH30023AP0G0500
LACH30023AP0G05001
LACH30023AP0G1000
Disulfid didraselný
Disulfid didraselný
Vzorec:
K₂O₅S₂ MW: 222.33 Bod tání: 190 Hustota: 1.2 g/cm³ (20 °C) |
MDL Number:
MFCD00167605 CAS číslo: 16731-55-8 EINECS: 240-795-3 Merck Index: 13,07729 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...