Dihydrogenfosforečnan draselný
Dodavatel: Lach-ner S.R.O.
Synonyms:
Monopotassiumphosphate, prim.-Potassium phosphate, Monopotassium phosphate, Potassium phosphate monobasic, Monobasic Potassium Phosphate, Potassium dihydrogen orthophosphate
30016CP0G0500
30016CP0G05001
LACH30016CP0G0500EA
255
CZK
LACH30016CP0G0500
LACH30016CP0G05001
Dihydrogenfosforečnan draselný
Dihydrogenfosforečnan draselný
Vzorec:
KH₂PO₄ Bod varu: ∼100 °C (1013 hPa) Bod tání: 253 °C Hustota: 2,3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011401 CAS číslo: 7778-77-0 EINECS: 231-913-4 REACH: 01-2119490224-41 Merck Index: 13,07744 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...