Tisk…

Tris-(hydroxymethyl)aminomethan

Dodavatel: Lach-ner S.R.O.
Varování

40159CP0G0250 40159CP0G0500 40159CP0G10001 40159CP0G01001 40159CP0G05001
LACH40159CP0G0250EA 718 CZK
LACH40159CP0G0250 LACH40159CP0G0500 LACH40159CP0G10001 LACH40159CP0G01001 LACH40159CP0G05001
Tris-(hydroxymethyl)aminomethan
Tris-(hydroxymethyl)aminomethan
Vzorec: NH₂C(CH₂OH)₃
Bod varu: 357 °C (1013 hPa)
Bod tání: 169…172 °C
Hustota: 1,328 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004679
CAS číslo: 77-86-1
EINECS: 201-064-4
Merck Index: 13,09842
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11a03d1ab6b2c11b9a2c83f0a73fc4264cec0110&vid=5ce890cdf298a59a7900a07fee1c857a5e5e61e0&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR