Hydrogenftalát draselný
Dodavatel: Lach-ner S.R.O.
Synonyms:
Potassium phthalate monobasic, Potassium biphthalate, Potassium Acid Phthalate, KHP, Phthalic Acid Monopotassium Salt, Phthalic acid potassium salt
40063AP0G1000
40063AP0G0500
40063AP0G01001
LACH40063AP0G1000EA
4720
CZK
LACH40063AP0G1000
LACH40063AP0G0500
LACH40063AP0G01001
Hydrogenftalát draselný
Hydrogenftalát draselný
Vzorec:
HOOCC₆H₄COOK MW: 204,22 g/mol Bod tání: 295…300 °C Hustota: 1,6362 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00013070 CAS číslo: 877-24-7 EINECS: 212-889-4 Merck Index: 13,07694 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...