Tisk…

3-Allylsalicylaldehyde, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3-AllylSalicylaldehyde, 3-Allyl-2-hydroxybenzaldehyde

M-Clarity™ quality level = MQ100

636339-1G 636339-5G
SIAL636339-1GEA 1050 CZK
SIAL636339-1G SIAL636339-5G
3-Allylsalicylaldehyde, Sigma-Aldrich®
3-Allylsalicylaldehyde
Vzorec: C₁₀H₁₀O₂
MW: 162,19 g/mol
MDL Number: MFCD00037364
CAS číslo: 24019-66-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR