3-Allylsalicylaldehyde 97
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
3-AllylSalicylaldehyde, 3-Allyl-2-hydroxybenzaldehyde
H53475.06
H53475.14
H53475.06EA
2910
CZK
H53475.06
H53475.14
3-Allylsalicylaldehyde 97
3-Allylsalicylaldehyde
Purity: 97%
CAS No.: 24019-66-7
Molecular Formula: C10H10O2
Formula Weight: 162.19
Flash Point: 107°(225°F)
Refractive Index: 1.5645
MDL No.: MFCD00037364
Vzorec:
C₁₀H₁₀O₂ MW: 162.19 g/mol Hustota: 1.099 Bod vzplanutí: 107 °C (225 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00037364 CAS číslo: 24019-66-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...