trans-2-Butenyl acetate 95%
Dodavatel: Thermo Scientific
|
Varování
|
L09734.06
L09734.14
L09734.06EA
2450
CZK
L09734.06
L09734.14
trans-2-Butenyl acetate 95%
trans-2-Butenyl acetate
Purity: 95%
CAS No.: 628-08-0
Molecular Formula: C6H10O2
Formula Weight: 114.15
Flash Point: 31°(87°F)
Refractive Index: 1.4200
Boiling Point: 128-130°
Beilstein Reference: 1720994
UN No.: 3272
MDL No.: MFCD00039912
Vzorec:
C₆H₁₀O₂ MW: 114,15 g/mol Bod varu: 128…130 °C Hustota: 0,919 Bod vzplanutí: 31 °C (87 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00039912 CAS číslo: 628-08-0 UN: 3272 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...