5-Hexenyl acetate ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
H53445.06
H53445.14
H53445.22
H53445.06EA
1370
CZK
H53445.06
H53445.14
H53445.22
5-Hexenyl acetate ≥97%
5-Hexenyl acetate
Purity: 97%
CAS No.: 5048-26-0
Molecular Formula: C8H14O2
Formula Weight: 142.20
Flash Point: 60°(142°F)
Refractive Index: 1.423
Boiling Point: 173-174°
UN No.: 3272
MDL No.: MFCD00060856
Vzorec:
C₈H₁₄O₂ MW: 142.20 g/mol Bod varu: 173…174 °C Hustota: 0.883 Bod vzplanutí: 60 °C (142 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00060856 CAS číslo: 5048-26-0 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...