Tisk…

L(-)-Bornyl acetate 95%

Dodavatel: Thermo Fisher Scientific
Varování

B22957.14 B22957.22 B22957.36
B22957.14EA 827 CZK
B22957.14 B22957.22 B22957.36
L(-)-Bornyl acetate 95%
L(-)-Bornyl acetate

Purity: 95% CAS No.: 5655-61-8 Molecular Formula: C12H20O2 Formula Weight: 196.29 Flash Point: 84°(184°F) Refractive Index: 1.4630 Boiling Point: 223-224° Beilstein Reference: 2502036 Merck Reference: 14,1339 MDL No.: MFCD00135942

Vzorec: C₁₂H₂₀O₂
MW: 196.29 g/mol
Bod varu: 223…224 °C
Hustota: 0.980
Bod vzplanutí: 84 °C (184 °F)
Storage Temperature: Ambient
MDL Number: MFCD00867808
CAS číslo: 5655-61-8
EINECS: 227-101-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR