Tisk…

Diethyl (3-chloropropyl)malonate ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Diethyl 2-(3-chloropropyl)malonate

L14776.06 L14776.14
L14776.06EA 1270 CZK
L14776.06 L14776.14
Diethyl (3-chloropropyl)malonate ≥98%
Diethyl (3-chloropropyl)malonate

Purity: 98% CAS No.: 18719-43-2 Molecular Formula: C10H17ClO4 Formula Weight: 236.70 Flash Point: 110°(230°F) Refractive Index: 1.4430 Boiling Point: 102°/1mm Beilstein Reference: 1785711 MDL No.: MFCD00009169

Vzorec: C₁₀H₁₇ClO₄
Bod varu: 102 °C (1 mmHg)
Hustota: 1,101
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009169
CAS číslo: 18719-43-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR