5-Amino-1-methyl-3-phenyl-1H-pyrazole 97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
1-Methyl-3-phenyl-1H-pyrazol-5-amine
A15754.03
A15754.06
A15754.03EA
725
CZK
A15754.03
A15754.06
5-Amino-1-methyl-3-phenyl-1H-pyrazole 97%
5-Amino-1-methyl-3-phenyl-1H-pyrazole
Purity: 97%
CAS No.: 10199-50-5
Molecular Formula: C10H11N3
Formula Weight: 173.22
Odour: Odourless
Melting Point: 124-130°
Beilstein Reference: 743807
MDL No.: MFCD00067874
Vzorec:
C₁₀H₁₁N₃ MW: 173,22 g/mol Bod tání: 124…130 °C Storage Temperature: Ambient |
MDL Number:
MFCD00067874 CAS číslo: 10199-50-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...