Tisk…

Ethyl isocyanatoacetate ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

L10609.06 L10609.14 L10609.03
L10609.06EA 2110 CZK
L10609.06 L10609.14 L10609.03
Ethyl isocyanatoacetate ≥98%
Ethyl isocyanatoacetate

Purity: 98% CAS No.: 2949-22-6 Molecular Formula: C5H7NO3 Formula Weight: 129.12 Storage Temperature: 2°C to 8°C Flash Point: 72°(161°F) Refractive Index: 1.4240 Odour: Pungent Boiling Point: 66-68°/11mm Beilstein Reference: 970503 MDL No.: MFCD00002040

Vzorec: C₅H₇NO₃
MW: 129.12 g/mol
Bod varu: 66…68 °C (11 mmHg)
Hustota: 1.151
Bod vzplanutí: 72 °C (161 °F)
MDL Number: MFCD00002040
CAS číslo: 2949-22-6
EINECS: 220-967-4
UN: 2206
ADR: 6.1 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR