Tisk…

L(+)-Lyxose ≥99%

Dodavatel: Thermo Scientific

B21583.03 B21583.06
B21583.03EA 3060 CZK
B21583.03 B21583.06
L(+)-Lyxose ≥99%
L(+)-Lyxose

Purity: 99% CAS No.: 1949-78-6 Molecular Formula: C5H10O5 Formula Weight: 150.13 Melting Point: 108-112° MDL No.: MFCD00064111

Vzorec: C₅H₁₀O₅
MW: 150,13 g/mol
Bod tání: 108…112 °C
Storage Temperature: Ambient
MDL Number: MFCD00064111
CAS číslo: 1949-78-6
EINECS: 217-763-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR