Tisk…

D-(-)-Lyxose mixture of anomers 99+%

Dodavatel: Thermo Scientific

205230050 205230250
ACRO205230050EA 3300 CZK
ACRO205230050 ACRO205230250
D-(-)-Lyxose mixture of anomers 99+%
D-(-)-Lyxose
Vzorec: C₅H₁₀O₅
MW: 150,13 g/mol
Bod tání: 108…112 °C
Storage Temperature: Ambient
MDL Number: MFCD00064362
CAS číslo: 1114-34-7
EINECS: 214-212-8

Objednat

Výsledky specifikačního testu

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
HPLC ≥99.0 % (ELSD)
Specific optical rotation -13° to -15° (20°C, 589 nm) (c=2, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR