1-(Methylsulphonyl)piperazine
Dodavatel: Apollo Scientific
Synonyms:
1-Methanesulphonylpiperazine
OR0841-5G
OR0841-25G
OR0841-100G
APOSOR0841-5GEA
662
CZK
APOSOR0841-5G
APOSOR0841-25G
APOSOR0841-100G
1-(Methylsulphonyl)piperazine
1-(Methylsulphonyl)piperazine
Vzorec:
C₅H₁₂N₂O₂S MW: 164,23 g/mol |
MDL Number:
MFCD02153225 CAS číslo: 55276-43-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...