Tisk…

2-Nitrophenol-[2,3,5,6-D₄] (98% D)

Dodavatel: LGC Standards PROMOCHEM

CILDLM-295-0.1 CILDLM2950.25
PROOCILDLM-295-0.1EA 8720 CZK
PROOCILDLM-295-0.1 PROOCILDLM2950.25
2-Nitrophenol-[2,3,5,6-D₄] (98% D)
2-Nitrophenol-[2,3,4,5-D₄]
Vzorec: C₆HD₄NO₃
CAS číslo: 93951-78-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR