D-(+)-Glucose-[2-¹³C] Isotope_(99% 13C), Sigma-Aldrich®
Dodavatel: Merck
MQ200
310794-1G
310794-250MG
SIAL310794-1GEA
32630
CZK
SIAL310794-1G
SIAL310794-250MG
D-(+)-Glucose-[2-¹³C] Isotope_(99% 13C), Sigma-Aldrich®
D-(+)-Glucose-[2-¹³C]
Vzorec:
C₆H₁₂O₆ |
MDL Number:
MFCD00037546 CAS číslo: 105931-74-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...