Tisk…

(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP), Sigma-Aldrich®

Dodavatel: Merck

Synonyms: PyBOP, ((1H-Benzo[d][1,2,3]triazol-1-yl)oxy)tri(pyrrolidin-1-yl)phosphonium hexafluorophosphate(V), 1H-Benzotriazol-1-yloxytri(1-pyrrolidinyl)phosphonium hexafluorophosphate

M-Clarity™ quality level = MQ200

12805-1G-F 12805-25G-F 12805-5G-F
SIAL12805-1G-FEA 1450 CZK
SIAL12805-1G-F SIAL12805-25G-F SIAL12805-5G-F
(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP), Sigma-Aldrich®
(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP)
Vzorec: C₁₈H₂₈F₆N₆OP₂
MW: 520.4 g/mol
MDL Number: MFCD00077411
CAS číslo: 128625-52-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR