Tisk…

4-Benzyloxy-2,6-dimethoxybenzaldehyde polymer-supported 1,0-1,5 mmol/g on resin, 1% cross-linked 100-200 mesh, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

538213-25G 538213-5G
SIAL538213-25GEA 30650 CZK
SIAL538213-25G SIAL538213-5G
4-Benzyloxy-2,6-dimethoxybenzaldehyde polymer-supported 1,0-1,5 mmol/g on resin, 1% cross-linked 100-200 mesh, Sigma-Aldrich®
4-Benzyloxy-2,6-dimethoxybenzaldehyde
Vzorec: C₁₆H₁₆O₄
CAS číslo: 679428-14-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR