Tisk…

Estradiol, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

E2758-1G E2758-250MG E2758-5G
SIALE2758-1GEA 3630 CZK
SIALE2758-1G SIALE2758-250MG SIALE2758-5G
Estradiol, Sigma-Aldrich®
Estradiol
Vzorec: C₁₈H₂₄O₂
MDL Number: MFCD00003693
CAS číslo: 50-28-2
EINECS: 200-023-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR