Estradiol, (max. 3% H₂O) 99% (dry weight)
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
beta-Estradiol, 17β-Estradiol
L03801.03
L03801.06
L03801.14
L03801.03EA
1780
CZK
L03801.03
L03801.06
L03801.14
Estradiol, (max. 3% H₂O) 99% (dry weight)
Estradiol
Purity: 99%
CAS No.: 50-28-2
Molecular Formula: C18H24O2
Formula Weight: 272.39
Melting Point: 175-178°
Beilstein Reference: 1914275
Merck Reference: 14,3703
Optical Rotation: +79° (c=1 in dioxane)
MDL No.: MFCD00003693
Vzorec:
C₁₈H₂₄O₂ MW: 272.39 g/mol Bod tání: 175…178 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003693 CAS číslo: 50-28-2 EINECS: 200-023-8 Merck Index: 14,3703 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...