2-Methoxyestradiol, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
(8R,9S,13S,14S,17S)-2-Methoxy-13-methyl-7,8,9,11,12,13,14,15,16,17-decahydro-6H-cyclopenta[a]phenanthrene-3,17-diol
M-Clarity™ quality level = MQ200
M6383-10MG
M6383-50MG
M6383-5MG
SIALM6383-10MGEA
3770
CZK
SIALM6383-10MG
SIALM6383-50MG
SIALM6383-5MG
2-Methoxyestradiol, Sigma-Aldrich®
2-Methoxyestradiol
Vzorec:
C₁₉H₂₆O₃ MW: 302,408 g/mol |
MDL Number:
MFCD00010489 CAS číslo: 362-07-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...