2-Methoxyestradiol
Dodavatel: Cayman Chemical
Synonyms:
(8R,9S,13S,14S,17S)-2-Methoxy-13-methyl-7,8,9,11,12,13,14,15,16,17-decahydro-6H-cyclopenta[a]phenanthrene-3,17-diol
13021-5
13021-10
13021-25
13021-50
CAYM13021-5EA
1290
CZK
CAYM13021-5
CAYM13021-10
CAYM13021-25
CAYM13021-50
2-Methoxyestradiol
2-Methoxyestradiol
Vzorec:
C₁₉H₂₆O₃ MW: 302.408 g/mol |
MDL Number:
MFCD00010489 CAS číslo: 362-07-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...