Tisk…

(R)-(-)-1-Boc-3-hydroxypyrrolidine ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: (R)-(-)-N-Boc-3-pyrrolidinol, tert-butyl (3R)-3-hydroxypyrrolidine-1-carboxylate, (R)-N-(tert-Butoxycarbonyl)-3-hydroxypyrrolidine

H27590.03 H27590.06
H27590.03EA 2420 CZK
H27590.03 H27590.06
(R)-(-)-1-Boc-3-hydroxypyrrolidine ≥98%
(R)-(-)-1-Boc-3-hydroxypyrrolidine

Purity: 98% CAS No.: 109431-87-0 Molecular Formula: C9H17NO3 Formula Weight: 187.24 Melting Point: 62-65° MDL No.: MFCD01317838

Vzorec: C₉H₁₇NO₃
MW: 187,24 g/mol
Bod tání: 62…65 °C
Storage Temperature: Ambient
MDL Number: MFCD01317838
CAS číslo: 109431-87-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR