Tisk…

(R)-(+)-1-Boc-3-aminopyrrolidine 97%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: (R)-1-Boc-3-aminopyrrolidine, (3R)-(+)-1-Boc-3-aminopyrrolidine, (3R)-(+)-3-Amino-1-Boc-pyrrolidine, (R)-tert-Butyl 3-aminopyrrolidine-1-carboxylate, (3R)-(+)-1-(tert-Butoxycarbonyl)-3-aminopyrrolidine, (3R)-(+)-3-Amino-1-(tert-butoxycarbonyl)pyrrolidine

H28188.06 H28188.03
H28188.06EA 12700 CZK
H28188.06 H28188.03
(R)-(+)-1-Boc-3-aminopyrrolidine 97%
(R)-(+)-1-Boc-3-aminopyrrolidine

Purity: 97% CAS No.: 147081-49-0 Molecular Formula: C9H18N2O2 Formula Weight: 186.25 Storage Temperature: 2°C to 8°C Flash Point: 91°(196°F) Refractive Index: 1.4720 Boiling Point: 243-244° Optical Rotation: +3.5° (c=1 in chloroform) UN No.: 2735 MDL No.: MFCD03419272

Vzorec: C₉H₁₈N₂O₂
MW: 186,25 g/mol
Bod varu: 243…244 °C
Hustota: 1,098
Bod vzplanutí: 91 °C (196 °F)
MDL Number: MFCD03419272
CAS číslo: 147081-49-0
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR