Tisk…

[2-(1,3-Dioxan-2-yl)ethyl]triphenylphosphonium bromide, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (2-(1,3-Dioxan-2-yl)ethyl)triphenylphosphonium bromide

M-Clarity™ quality level = MQ200

219592-25G 219592-5G
SIAL219592-25GEA 0 CZK
SIAL219592-25G SIAL219592-5G
[2-(1,3-Dioxan-2-yl)ethyl]triphenylphosphonium bromide, Sigma-Aldrich®
[2-(1,3-Dioxan-2-yl)ethyl]triphenylphosphonium bromide
Vzorec: C₂₄H₂₆BrO₂P
MW: 457,36 g/mol
MDL Number: MFCD00012000
CAS číslo: 69891-92-5
EINECS: 274-189-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR