Tisk…

Dodecacarbonyltriruthenium 99%

Dodavatel: Thermo Fisher Scientific

Synonyms: Triruthenium dodecacarbonyl, Ruthenium carbonyl

224000010 224000050
ACRO224000010EA 6030 CZK
ACRO224000010 ACRO224000050
Dodecacarbonyltriruthenium 99%
Dodecacarbonyltriruthenium
Vzorec: C₁₂O₁₂Ru₃
MW: 639.33 g/mol
Bod tání: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00011209
CAS číslo: 15243-33-1
EINECS: 239-287-4
ADR: 6.1 I

Objednat

Výsledky specifikačního testu

Appearance Orange Crystals
Infrared spectrum Conforms
Elemental Analysis ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR