Tisk…

Mineral oil, white (petroleum), 75 cSt

Dodavatel: LGC Standards PROMOCHEM

Synonyms: Matrix oil, Mineral oil, Paraffin oil

CON150-075-002 CON150-075-004 CON150-075-006
PROOCON150-075-002EA 3540 CZK
PROOCON150-075-002 PROOCON150-075-004 PROOCON150-075-006
Mineral oil, white (petroleum), 75 cSt
Mineral oil, white (petroleum)
MDL Number: MFCD00131611
CAS číslo: 8042-47-5
EINECS: 232-455-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR