Tisk…
Methylamin 33% in ethanol, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
534102-250ML 534102-1L
SIAL534102-250MLEA 2150 CZK
SIAL534102-250ML SIAL534102-1L
Methylamin 33% in ethanol, Sigma-Aldrich®
Methylamin
Vzorec: CH₅N
Storage Temperature: Ambient
MDL Number: MFCD00008104
CAS číslo: 74-89-5
EINECS: 200-820-0
UN: 2924
ADR: 3,II