3-Amino-2-cyclohexen-1-one, (max. 5% H₂O) 98% (dry weight)
Dodavatel: Thermo Scientific
|
Varování
|
A11474.06
A11474.14
A11474.06EA
766
CZK
A11474.06
A11474.14
3-Amino-2-cyclohexen-1-one, (max. 5% H₂O) 98% (dry weight)
3-Amino-2-cyclohexen-1-one
Purity: 98%
CAS No.: 5220-49-5
Molecular Formula: C6H9NO
Formula Weight: 111.15
Melting Point: 128-133°(dried)
Beilstein Reference: 471354
Vzorec:
C₆H₉NO MW: 111,14 g/mol Bod varu: 211 °C (1013 hPa) Bod tání: 128…133 °C (anhydr.) Storage Temperature: Ambient |
MDL Number:
MFCD00013783 CAS číslo: 5220-49-5 EINECS: 226-014-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...