(tert-Butoxycarbonylmethyl)triphenylphosphonium bromide, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
(2-(tert-Butoxy)-2-oxoethyl)triphenylphosphonium bromide
369047-50G
SIAL369047-50GEA
6410
CZK
SIAL369047-50G
(tert-Butoxycarbonylmethyl)triphenylphosphonium bromide, Sigma-Aldrich®
(tert-Butoxycarbonylmethyl)triphenylphosphonium bromide
Vzorec:
C₂₄H₂₆BrO₂P MW: 457.35 g/mol |
MDL Number:
MFCD00075527 CAS číslo: 59159-39-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...