2-(p-Tolyl)pyridine ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
H26574.14
H26574.06
H26574.14EA
5010
CZK
H26574.14
H26574.06
2-(p-Tolyl)pyridine ≥98%
2-(p-Tolyl)pyridine
Purity: 98%
CAS No.: 4467-06-5
Molecular Formula: C12H11N
Formula Weight: 169.22
Flash Point:
Vzorec:
C₁₂H₁₁N MW: 169.22 g/mol Bod varu: 170…180 °C (20 mmHg) Hustota: 0.99 Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006281 CAS číslo: 4467-06-5 EINECS: 224-734-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...