Tisk…

1,3-Di-tert-butyltoluene ≥98%

Dodavatel: Thermo Fisher Scientific

Synonyms: 1,3-Di-tert-butyl-5-methylbenzene

L11961.14 AB179366.0025
L11961.14EA 3340 CZK
L11961.14 ABCRAB179366.0025
1,3-Di-tert-butyltoluene ≥98%
1,3-Di-tert-butyltoluene

Purity: 98+% CAS No.: 15181-11-0 Molecular Formula: C15H24 Formula Weight: 204.36 Flash Point: 91°(195°F) Refractive Index: 1.4900 Melting Point: 31-32° Boiling Point: 243-244° Beilstein Reference: 2043032 MDL No.: MFCD00026300

Vzorec: C₁₅H₂₄
MW: 204.36 g/mol
Bod varu: 243…244 °C
Bod tání: 31…32 °C
Hustota: 0.860
Bod vzplanutí: 91 °C (195 °F)
Storage Temperature: Ambient
MDL Number: MFCD00026300
CAS číslo: 15181-11-0
EINECS: 239-230-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR