3-Phenoxyphenylboronic acid ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
(3-Phenoxyphenyl)boronic acid, 3-Phenoxybenzeneboronic acid
H32879.MD
H32879.03
H32879.MDEA
2130
CZK
H32879.MD
H32879.03
3-Phenoxyphenylboronic acid ≥97%
3-Phenoxyphenylboronic acid
Purity: 97+%
CAS No.: 221006-66-2
Molecular Formula: C12H11BO3
Formula Weight: 214.03
Melting Point: 138-143°
MDL No.: MFCD07780272
Vzorec:
C₁₂H₁₁BO₃ MW: 214,03 g/mol Bod tání: 138…143 °C Storage Temperature: Ambient |
MDL Number:
MFCD07780272 CAS číslo: 221006-66-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...