4-Amino-3-chlorobenzonitrile 98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
A12154.03
A12154.06
A12154.14
A12154.03EA
567
CZK
A12154.03
A12154.06
A12154.14
4-Amino-3-chlorobenzonitrile 98%
4-Amino-3-chlorobenzonitrile
Purity: 98%
CAS No.: 21803-75-8
Molecular Formula: C7H5ClN2
Formula Weight: 152.58
Melting Point: 98-104°
Beilstein Reference: 1817655
UN No.: 3439
MDL No.: MFCD00052917
Vzorec:
C₇H₅ClN₂ MW: 152.58 g/mol Bod tání: 98…104 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052917 CAS číslo: 21803-75-8 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...