1,1,1,3,3,3-Hexafluoroisopropyl methacrylate ≥99% stabilised
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Hexafluoroisopropyl methacrylate, 1,1,1,3,3,3-Hexafluoroisopropylmethacrylate
B21447.06
B21447.14
B21447.06EA
1770
CZK
B21447.06
B21447.14
1,1,1,3,3,3-Hexafluoroisopropyl methacrylate ≥99% stabilised
1,1,1,3,3,3-Hexafluoroisopropyl methacrylate
Stabilized
Purity: 99%
CAS No.: 3063-94-3
Molecular Formula: CH2=C(CH3)CO2CH(CF3)2
Formula Weight: 236.12
Flash Point: 14°(57°F)
Refractive Index: 1.3310
Boiling Point: 99°
UN No.: 1993
MDL No.: MFCD00040105
Vzorec:
C₇H₆F₆O₂ Bod varu: 99 °C Hustota: 1,302 Bod vzplanutí: 14 °C (57 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00040105 CAS číslo: 3063-94-3 EINECS: 221-309-9 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...