4-Bromo-2-chloro-6-methylaniline ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
L12590.06
L12590.14
L12590.06EA
1670
CZK
L12590.06
L12590.14
4-Bromo-2-chloro-6-methylaniline ≥98%
4-Bromo-2-chloro-6-methylaniline
Purity: 98%
CAS No.: 30273-42-8
Molecular Formula: C7H7BrClN
Formula Weight: 220.50
Melting Point: 37-40°
Beilstein Reference: 2717133
UN No.: 2811
MDL No.: MFCD00041432
Vzorec:
C₇H₇BrClN Bod tání: 37…40 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041432 CAS číslo: 30273-42-8 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...