(5-Carboxypentyl)triphenylphosphonium bromide ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
B20588.03
B20588.06
B20588.03EA
776
CZK
B20588.03
B20588.06
(5-Carboxypentyl)triphenylphosphonium bromide ≥97%
(5-Carboxypentyl)triphenylphosphonium bromide
Purity: 97%
CAS No.: 50889-29-7
Molecular Formula: HO2C(CH2)5P(C6H5)3Br
Formula Weight: 457.36
Melting Point: 200-202°
MDL No.: MFCD00055556
Vzorec:
C₂₄H₂₆BrO₂P MW: 457,36 g/mol Bod tání: 200…202 °C Storage Temperature: Ambient |
MDL Number:
MFCD00055556 CAS číslo: 50889-29-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...