Tisk…

(3-Carboxypropyl)triphenylphosphonium bromide, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3-Carboxypropyltriphenylphosphonium bromide

M-Clarity™ quality level = MQ100

349720-100G 349720-25G
SIAL349720-100GEA 7580 CZK
SIAL349720-100G SIAL349720-25G
(3-Carboxypropyl)triphenylphosphonium bromide, Sigma-Aldrich®
(3-Carboxypropyl)triphenylphosphonium bromide
Vzorec: C₂₂H₂₂BrO₂P
MW: 429,30 g/mol
MDL Number: MFCD00274196
CAS číslo: 17857-14-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR