Kaempferol, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
3,5,7-Trihydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one
M-Clarity™ quality level = MQ100
K0133-100MG
K0133-10MG
K0133-500MG
K0133-50MG
SIALK0133-100MGEA
29620
CZK
SIALK0133-100MG
SIALK0133-10MG
SIALK0133-500MG
SIALK0133-50MG
Kaempferol, Sigma-Aldrich®
Kaempferol
Vzorec:
C₁₅H₁₀O₆ MW: 286,24 g/mol |
MDL Number:
MFCD00016938 CAS číslo: 520-18-3 EINECS: 208-287-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...