Tisk…

L-(-)-Fucose, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (-)-Fucose, 6-Deoxy-L-Galactose, (2S,3R,4R,5S)-2,3,4,5-Tetrahydroxyhexanal

M-Clarity™ quality level = MQ200

F2252-5G F2252-100G F2252-10MG F2252-25G
SIALF2252-5GEA 9660 CZK
SIALF2252-5G SIALF2252-100G SIALF2252-10MG SIALF2252-25G
L-(-)-Fucose, Sigma-Aldrich®
L(-)-Fucose
Vzorec: C₆H₁₂O₅
MW: 164,16 g/mol
MDL Number: MFCD00069812
CAS číslo: 2438-80-4
EINECS: 219-452-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR