Tisk…

Geranyl propionate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ300

W251704-1KG-K W251704-5KG-K
SIALW251704-1KG-KEA 4100 CZK
SIALW251704-1KG-K SIALW251704-5KG-K
Geranyl propionate, Sigma-Aldrich®
Geranyl propionate
Vzorec: C₁₃H₂₂O₂
MDL Number: MFCD00027009
CAS číslo: 105-90-8
EINECS: 203-344-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR