Tisk…

Geranyl formate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3,7-Dimethylocta-2,6-dien-1-yl formate

M-Clarity™ quality level = MQ300

W251402-10KG-K W251402-1KG-K W251402-5KG-K
SIALW251402-10KG-KEA 20340 CZK
SIALW251402-10KG-K SIALW251402-1KG-K SIALW251402-5KG-K
Geranyl formate, Sigma-Aldrich®
Geranyl formate
Vzorec: C₁₁H₁₈O₂
MDL Number: MFCD00021047
CAS číslo: 105-86-2
EINECS: 203-339-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR