Trimethylamine-N-oxide dihydrate ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Trimethylamine oxide dihydrate
A14916.14
A14916.06
A14916.22
A14916.36
A14916.14EA
1440
CZK
A14916.14
A14916.06
A14916.22
A14916.36
Trimethylamine-N-oxide dihydrate ≥98%
Trimethylamine-N-oxide dihydrate
Purity: 98+%
CAS No.: 62637-93-8
Molecular Formula: (CH3)3NO.2H2O
Formula Weight: 111.14 (75.11anhy)
Melting Point: 96-101°
Beilstein Reference: 3612927
Merck Reference: 14,9710
MDL No.: MFCD00149077
Vzorec:
C₃H₉NO·2H₂O MW: 111.14 g/mol (75.11 anhy) Bod tání: 96…101 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149077 CAS číslo: 62637-93-8 EINECS: 214-675-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...