D-(+)-Turanose ≥98%
Dodavatel: Thermo Scientific
Synonyms:
(3S,4R,5R)-1,4,5,6-Tetrahydroxy-3-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexan-2-one, Turanose
B21224.03
B21224.06
B21224.03EA
2080
CZK
B21224.03
B21224.06
D-(+)-Turanose ≥98%
D-(+)-Turanose
Purity: 98%
CAS No.: 547-25-1
Molecular Formula: C12H22O11
Formula Weight: 342.30
Melting Point: ca 170° dec.
Beilstein Reference: 93771
Merck Reference: 14,9821
MDL No.: MFCD00006606
Vzorec:
C₁₂H₂₂O₁₁ MW: 342,30 g/mol Bod tání: ∼ 170 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00006606 CAS číslo: 547-25-1 EINECS: 208-918-5 Merck Index: 14,9821 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...