D-(+)-Cellobiose, Millipore®
Dodavatel: Merck
Synonyms:
β-D-Glc-(1→4)-D-Glc, 4-β-D-glucopyranosyl-D-glueopyranose, Cellose, β-Cellobiose, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal, 4-O-β-D-Glucopyranosyl-D-glucopyranose
M-Clarity™ quality level = MQ200
22150-10G
22150-250G
22150-50G
SIAL22150-10GEA
0
CZK
SIAL22150-10G
SIAL22150-250G
SIAL22150-50G
D-(+)-Cellobiose, Millipore®
D-(+)-Cellobiose
Vzorec:
C₁₂H₂₂O₁₁ MW: 342,30 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00136034 CAS číslo: 528-50-7 EINECS: 208-436-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...