Benzyloxyacetaldehyde diethyl acetal ≥98%
Dodavatel: Thermo Fisher Scientific
Varování
|
A14474.09
A14474.18
A14474.30
A14474.09EA
2520
CZK
A14474.09
A14474.18
A14474.30
Benzyloxyacetaldehyde diethyl acetal ≥98%
Benzyloxyacetaldehyde diethyl acetal
Purity: 98%
CAS No.: 42783-78-8
Molecular Formula: C6H5CH2OCH2CH(OCH2CH3)2
Formula Weight: 224.30
Flash Point: 79°(174°F)
Refractive Index: 1.4790
Boiling Point: 99-100°/0.6mm
Beilstein Reference: 3278795
MDL No.: MFCD00051786
Vzorec:
C₁₃H₂₀O₃ MW: 224.30 g/mol Bod varu: 99…100 °C (0.6 mmHg) Hustota: 0.987 Bod vzplanutí: 79 °C (174 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00051786 CAS číslo: 42783-78-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...