Tisk…

Potassium hexafluorozirconate ≥99%

Dodavatel: Thermo Scientific
Nebezpečí

86102.36 86102.A3
86102.36EA 3020 CZK
86102.36 86102.A3
Potassium hexafluorozirconate ≥99%
Potassium hexafluorozirconate

Purity: 99% CAS No.: 16923-95-8 Molecular Formula: K2ZrF6 Formula Weight: 283.41 Physical Form: -10 Mesh Density: 3.48 Solubility: Slightly soluble in cold water, soluble in hot water Merck Reference: 14,7639 UN No.: 3288 MDL No.: MFCD00042547


For manufacture of metallic zirconium

Vzorec: F₆K₂Zr
MW: 283,41 g/mol
Hustota: 3,48
Storage Temperature: Ambient
MDL Number: MFCD00042547
CAS číslo: 16923-95-8
EINECS: 240-985-6
UN: 3288
ADR: 6.1,III
Merck Index: 14,7639

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR