Nonylphenol (mixture of isomers), Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
290858-1L
290858-25ML
290858-3L
SIAL290858-1LEA
2650
CZK
SIAL290858-1L
SIAL290858-25ML
SIAL290858-3L
Nonylphenol (mixture of isomers), Sigma-Aldrich®
Nonylphenol (mixture of isomers)
Vzorec:
C₁₅H₂₄O |
MDL Number:
MFCD00002396 CAS číslo: 84852-15-3 EINECS: 284-325-5 UN: 3145 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...